Learn More
Aviva Systems Biology Corp KRT13 Antibody - C-terminal region (ARP42031_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42031_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against KRT13. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human KRT13. Alternate names: CK13, K13, MGC161462, MGC3781ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-58086