Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 3861MSM3P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: KRT14/532. Clonality: Monoclonal. Host: Mouse. Isotype: IgG3. Positive Control: A431 cells. Skin, Tonsil, Pro. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant full-length human KRT14 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 50kDa.Chromosome: 17q21.2. Gene Symbol: KRT14. Gene ID: 3861. Ensemble: ENSG00000186847. Uniprot: NP_000517.
Catalog No. 50-188-4886