Learn More
Aviva Systems Biology Corp KRT14 Antibody - C-terminal region (AVARP15002_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP15002_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against KRT14. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human KRT14. Alternate names: K14, NFJ, CK14, EBS3, EBS4ApplicationWB. Species reactivity: Guinea Pig, Mouse, Rabbit, Rat
Catalog No. 50-014-62645