Learn More
Aviva Systems Biology Corp KRT15 Antibody - C-terminal region (AVARP00005_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP00005_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KRT15. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human KRT15. Alternate names: CK15, K15, K1COApplicationIHC, WB. Species reactivity: Cow, Human, Mouse
Catalog No. 50-014-62443