Learn More
Aviva Systems Biology Corp KRT16 Antibody - middle region (ARP42240_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42240_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KRT16. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KRT16. Alternate names: CK16, K16, K1CP, KRT16A, NEPPK, FNEPPKApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-58160