Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 3872MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: KRT17/778. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2b kappa. Positive Control: T24 cells or Skin. Bladder or Cervix. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant full-length human KRT17 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 46kDa.Chromosome: 17q21.2. Gene Symbol: KRT17. Gene ID: 3872. Ensemble: ENSG00000128422. Uniprot: NP_000413.
Catalog No. 50-188-4892