Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 3875MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: WB. Additionally, this antibody has been validated by MS-QAVA(TM). Specificity has been quantitatively determined via Mass Spec. Conjugate/Tag: Unconjugated. Clone: DC10. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1. Positive Control: MCF-7, HeLa, A431 cells or Breast Cancer. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: PMC-42 human breast carcinoma cells. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 45kDa.Chromosome: 12q13.13. Gene Symbol: KRT18. Gene ID: 3875. Ensemble: ENSG00000111057. Uniprot: NP_000215.
Catalog No. 50-188-4894