Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 3875MSM7P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: C-04; same as Ks18.04. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: MCF-7, HeLa cells, Breast Cancer. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Cytoskeletal preparation extracted from the human vulvar squamous carcinoma cell line A431. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 45kDa.Chromosome: 12q13.13. Gene Symbol: KRT18. Gene ID: 3875. Ensemble: ENSG00000111057. Uniprot: NP_000215.
Catalog No. 50-188-4901