Learn More
Aviva Systems Biology Corp KRT18 Antibody - N-terminal region (ARP40205_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40205_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against KRT18. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human KRT18. Alternate names: K18, CYK18ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57339