Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC KRT18 Human HEK293 50ug
SDP

Catalog No. 5000196325
Encompass_Preferred

Recombinant human keratin 18 protein produced in HEK293 cells, supplied lyophilized with an N-terminal His affinity tag for use in antibody validation, biochemical assays, and cell biology studies.

  • Recombinant human keratin 18 fragment expressed in HEK293 cells.
  • Contains an N-terminal His affinity tag to facilitate purification.
  • High purity (>90%) as determined by reducing SDS-PAGE.
  • Low endotoxin level suitable for sensitive in vitro assays.
  • Lyophilized formulation with trehalose for enhanced stability.
  • Recommended storage at -20°C and aliquot freezing for extended shelf life.
  • Reconstitution recommended at ≥100 μg/mL in ddH₂O for optimal performance.

Catalog No. 50-001-96325 Supplier Medchemexpress LLC Supplier No. HYP7166850UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human keratin 18 protein produced in HEK293 cells, supplied lyophilized with an N-terminal His affinity tag for use in antibody validation, biochemical assays, and cell biology studies.

  • Recombinant human keratin 18 fragment expressed in HEK293 cells.
  • Contains an N-terminal His affinity tag to facilitate purification.
  • High purity (>90%) as determined by reducing SDS-PAGE.
  • Low endotoxin level suitable for sensitive in vitro assays.
  • Lyophilized formulation with trehalose for enhanced stability.
  • Recommended storage at -20°C and aliquot freezing for extended shelf life.
  • Reconstitution recommended at ≥100 μg/mL in ddH₂O for optimal performance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.