Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC KRT18 Human HEK293 5ug
SDP

Catalog No. 5000196731
Encompass_Preferred

Recombinant human keratin 18 (KRT18) expressed in HEK293 cells and supplied with an N-terminal 10xHis affinity tag. The protein is purified to >90% by reducing SDS-PAGE, has an approximate molecular weight of 50.8 kDa corresponding to residues S2-H430, and is provided in multiple small unit sizes suitable for biochemical assays, antibody validation, and protein-protein interaction studies.

  • Recombinant human keratin 18 expressed in HEK293 cells.
  • N-terminal 10xHis tag for affinity purification and detection.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Approximate molecular weight 50.8 kDa (S2-H430).
  • Available in small unit sizes for assay development and controls.
  • Suitable for biochemical assays, antibody validation, and interaction studies.

Catalog No. 50-001-96731 Supplier Medchemexpress LLC Supplier No. HYP716685UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human keratin 18 (KRT18) expressed in HEK293 cells and supplied with an N-terminal 10xHis affinity tag. The protein is purified to >90% by reducing SDS-PAGE, has an approximate molecular weight of 50.8 kDa corresponding to residues S2-H430, and is provided in multiple small unit sizes suitable for biochemical assays, antibody validation, and protein-protein interaction studies.

  • Recombinant human keratin 18 expressed in HEK293 cells.
  • N-terminal 10xHis tag for affinity purification and detection.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Approximate molecular weight 50.8 kDa (S2-H430).
  • Available in small unit sizes for assay development and controls.
  • Suitable for biochemical assays, antibody validation, and interaction studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.