Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 3880MSM7P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: KRT19/799 + KRT19/800. Clonality: Monoclonal. Host: Mouse. Isotype: IIgG2a kappa. Positive Control: MCF-7, HeLa cells, Breast Cancer. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant full-length human KRT19 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 40kDa.Chromosome: 17q21.2. Gene Symbol: KRT19. Gene ID: 3880. Ensemble: ENSG00000171345. Uniprot: NP_002267.
Catalog No. 50-188-4908