Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 3880MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Additionally, this antibody has been validated by MS-QAVA(TM). Specificity has been quantitatively determined via Mass Spec. Conjugate/Tag: Unconjugated. Clone: A53-B/A2.26. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2a kappa. Positive Control: MCF-7, HeLa, Hep-G2 cells, Breast Cancer. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Human breast cancer MCF-7 cells. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 40kDa.Chromosome: 17q21.2. Gene Symbol: KRT19. Gene ID: 3880. Ensemble: ENSG00000171345. Uniprot: NP_002267.
Catalog No. 50-188-4904