Learn More
Aviva Systems Biology Corp KRT20 Antibody - middle region (ARP42529_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42529_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against KRT20. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KRT20. Alternate names: CD20, CK20, K20, KRT21, MGC35423, CK-20ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58256