Learn More
Aviva Systems Biology Corp KRT222P Antibody - N-terminal region (ARP52914_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52914_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KRT222P. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KRT222P. Alternate names: KA21, MGC45562, KRT222PApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60869