Learn More
Aviva Systems Biology Corp KRT23 Antibody - middle region (ARP47466_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47466_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KRT23. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KRT23. Alternate names: CK23, DKFZP434G032, HAIK1, K23, MGC26158ApplicationIHC, WB. Species reactivity: Dog, Guinea Pig, Horse, Human
Catalog No. 50-014-59557