Learn More
Aviva Systems Biology Corp KRT24 Antibody - middle region (ARP52386_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52386_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KRT24. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KRT24. Alternate names: FLJ20261, MGC138169, MGC138173, K24, KA24ApplicationWB. Species reactivity: Horse, Human, Pig
Catalog No. 50-014-60723