Learn More
Aviva Systems Biology Corp KRT2A Antibody - middle region (ARP41735_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41735_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KRT2A. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human KRT2A. Alternate names: KRT2E, KRTE, K2e, CK-2e, KRT2AApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53313