Learn More
Aviva Systems Biology Corp KRT36 Antibody - N-terminal region (ARP61214_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61214_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KRT36. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human KRT36. Alternate names: HA6, KRTHA6, hHa6ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-287-298