Learn More
Aviva Systems Biology Corp KRT4 Antibody - N-terminal region (ARP64964_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64964_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KRT4. It was validated on Western Blot. . Alternate names: CK4, CYK4, FLJ31692, K4, CK-4ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-288-403