Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents MSM13X948P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, IHC. Conjugate/Tag: Unconjugated. Clone: SPM268. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: HepG2 or A431 cells. Skin or Squamous cell carcinoma. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS, pH 7.4 with 0.05% BSA & 0.05% sodium azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Cytoskeletal preparation from HeLa cells. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 58kDa (CK5); 52.5kDa (CK8).Chromosome: 12q13.13 (CK5) & 17q21.2 (CK8). Gene Symbol: KRT5 & KRT8. Gene ID: 3856. Ensemble: ENSG00000170421. Uniprot: NP_001243222.
Catalog No. 50-188-5412