Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 3853MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: KRT6/1702. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2a kappa. Positive Control: Tonsil or basal cell carcinoma. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human full-length keratin 6 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 56kDa.Chromosome: 12q13.13 ;12q13.13 ;12q13.13. Gene Symbol: KRT6A ;KRT6B ;KRT6C. Gene ID: 286887. Ensemble: ENSG00000170465. Uniprot: NP_775109.
Catalog No. 50-188-4856