Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 3855MSM8P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: KRT7/1198. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1. Positive Control: HeLa cells. Carcinoma of ovary, lung, cervix, or breast. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant full-length human KRT7 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 55kDa.Chromosome: 12q13.13. Gene Symbol: KRT7. Gene ID: 3855. Ensemble: ENSG00000135480. Uniprot: NP_011536627.
Catalog No. 50-188-4865