Learn More
Aviva Systems Biology Corp KRT71 Antibody - middle region (ARP65462_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65462_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KRT71. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of Human KRT71. Alternate names: K6IRS1, KRT6IRS, KRT6IRS1, MGC119390, MGC119391ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-288-502