Learn More
Aviva Systems Biology Corp KRT75 Antibody - N-terminal region (ARP58768_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58768_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KRT75. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KRT75. Alternate names: K6HF, PFB, KB18ApplicationWB. Species reactivity: Cow, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-62418