Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents MSM241377P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: KRTL/1377. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1. Positive Control: Skin, Squamous cell carcinoma (SCC). Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human KRT77 protein fragment. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: the usefulness of keratins as markers in cancer research and tumor diagnosis.Chromosome: 17q21.2. Gene Symbol: KRT77. Gene ID: 374454. Ensemble: ENSG00000189182. Uniprot: XP_011536590.
Catalog No. 50-188-5452