Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents MSM4X253P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, IHC. Conjugate/Tag: Unconjugated. Clone: SPM115. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Skin, Squamous cell carcinoma (SCC). Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Human epidermal keratin. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 56.5kDa (CK10); 50kDa (CK14); 50kDa (CK15); 48kDa (CK16); 40kDa (CK19).Chromosome: 17q21.2. Gene Symbol: KRT77. Gene ID: 374454. Ensemble: ENSG00000189182. Uniprot: XP_011536590.
Catalog No. 50-188-5472