Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents MSM6691P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, IHC. Conjugate/Tag: Unconjugated. Clone: K8.8 + DC10. Clonality: Monoclonal. Host: Mouse. Isotype: IgG. Positive Control: MCF-7 or A431 cells. Skin, Colon, lung or breast carcinoma. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Keratin preparation from a human carcinoma (K8.8); PMC-42 human breast carcinoma cells (DC10). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 52.5kDa (CK8); 45kDa (CK18).Chromosome: 17q21.2 (CK8); 12q13.13 (CK18). Gene Symbol: KRT8 & KRT18. Gene ID: 3875. Ensemble: ENSG00000111057. Uniprot: NP_000215.
Catalog No. 50-188-5476