Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents MSM7770P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, IHC. Additionally, this antibody has been validated by MS-QAVA(TM). Specificity has been quantitatively de. Conjugate/Tag: Unconjugated. Clone: C-43 + DC10. Clonality: Monoclonal. Host: Mouse. Isotype: IgG. Positive Control: MCF-7 or A431 cells. Skin, Colon, lung or breast carcinoma. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Keratin preparation from a human carcinoma (C-43); PMC-42 human breast carcinoma cells (DC10). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C.
Catalog No. 50-188-5478