Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents MSM271346P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Additionally, this antibody has been validated by MS-QAVA(TM). Specificity has been quantitatively determined via Mass Spec. Conjugate/Tag: Unconjugated. Clone: KRT8.18/1346. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: MCF-7 or A431 cells. Skin, Colon, lung or breast carcinoma. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Cytoskeleton preparation from HeLa cells. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C.
Catalog No. 50-188-5455