Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 3856MSM3P1
Encompass_Preferred
Product Type: Antibody. Application: WB. Additionally, this antibody has been validated by MS-QAVA(TM). Specificity has been quantitatively determined via Mass Spec. Conjugate/Tag: Unconjugated. Clone: K8/383. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1. Positive Control: MCF-7 or A431 cells. Skin, colon, lung or breast carcinoma. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human full-length cytokeratin 8 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 52.5kDa.Chromosome: 12q13.13. Gene Symbol: KRT8. Gene ID: 3856. Ensemble: ENSG00000170421. Uniprot: NP_001243222.
Catalog No. 50-188-4870