Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 3856MSM1XP1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: SPM538. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1. Positive Control: MCF-7 or A431 cells. Skin, Colon, lung or breast carcinoma. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Cytoskeleton preparation containing cytokeratin 8. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 52.5kDa.Chromosome: 12q13.13. Gene Symbol: KRT8. Gene ID: 3856. Ensemble: ENSG00000170421. Uniprot: NP_001243222.
Catalog No. 50-188-4869