Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC KRT8 Human sf9 Hi 50ug
SDP

Catalog No. 5000195614
Encompass_Preferred

Recombinant human keratin 8 (KRT8) produced in Sf9 insect cells and supplied as a lyophilized protein with N-terminal His and C-terminal Myc tags. The preparation is low endotoxin (<1 EU/μg) and is lyophilized from PBS with trehalose to enhance stability. Reconstitute at ≥100 μg/mL in ddH2O. Store lyophilized at -20°C and aliquot frozen samples for extended storage.

  • Expression host: Sf9 insect cells
  • Affinity tags: His and Myc
  • Format: Lyophilized powder for enhanced stability
  • Endotoxin: <1 EU/μg (LAL)
  • Reconstitution recommendation: not less than 100 μg/mL in ddH2O
  • Storage: store lyophilized at -20°C; aliquot and freeze for long-term storage
  • Molecular weight: approximately 57.6 kDa

Catalog No. 50-001-95614 Supplier Medchemexpress LLC Supplier No. HYP7205250UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human keratin 8 (KRT8) produced in Sf9 insect cells and supplied as a lyophilized protein with N-terminal His and C-terminal Myc tags. The preparation is low endotoxin (<1 EU/μg) and is lyophilized from PBS with trehalose to enhance stability. Reconstitute at ≥100 μg/mL in ddH2O. Store lyophilized at -20°C and aliquot frozen samples for extended storage.

  • Expression host: Sf9 insect cells
  • Affinity tags: His and Myc
  • Format: Lyophilized powder for enhanced stability
  • Endotoxin: <1 EU/μg (LAL)
  • Reconstitution recommendation: not less than 100 μg/mL in ddH2O
  • Storage: store lyophilized at -20°C; aliquot and freeze for long-term storage
  • Molecular weight: approximately 57.6 kDa

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.