Learn More
Medchemexpress LLC KRT8 Human sf9 Hi 5ug
SDP
Catalog No. 5000195616
Encompass_Preferred
Recombinant human keratin 8 (KRT8) produced in Sf9 insect cells and supplied as a lyophilized powder with His and Myc affinity tags. The protein is formulated in PBS with 6% trehalose, has an approximate molecular weight of 57.6 kDa, and an endotoxin level <1 EU/μg. It is for research use and should be stored frozen; reconstitute to a concentration of at least 100 μg/mL for use.
  • Recombinant human protein expressed in Sf9 insect cells.
  • N-terminal His and C-terminal Myc tags for detection and purification.
  • Lyophilized formulation stabilized in PBS with 6% trehalose.
  • Low endotoxin level suitable for sensitive assays.
  • Recommended reconstitution concentration of at least 100 μg/mL.
  • Store frozen and aliquot for long-term stability.
Catalog No. 50-001-95616 Supplier Medchemexpress LLC Supplier No. HYP720525UG
Recombinant human keratin 8 (KRT8) produced in Sf9 insect cells and supplied as a lyophilized powder with His and Myc affinity tags. The protein is formulated in PBS with 6% trehalose, has an approximate molecular weight of 57.6 kDa, and an endotoxin level <1 EU/μg. It is for research use and should be stored frozen; reconstitute to a concentration of at least 100 μg/mL for use.
  • Recombinant human protein expressed in Sf9 insect cells.
  • N-terminal His and C-terminal Myc tags for detection and purification.
  • Lyophilized formulation stabilized in PBS with 6% trehalose.
  • Low endotoxin level suitable for sensitive assays.
  • Recommended reconstitution concentration of at least 100 μg/mL.
  • Store frozen and aliquot for long-term stability.