Learn More
Aviva Systems Biology Corp KRT9 Antibody - C-terminal region (ARP64510_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64510_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KRT9. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human KRT9. Alternate names: CK-9, EPPK, K9ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-288-278