Learn More
Aviva Systems Biology Corp KRTAP1-5 Antibody - C-terminal region (ARP60599_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60599_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KRTAP1-5. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human KRTAP1-5. Alternate names: KAP1.5, KRTAP1.5, MGC126604ApplicationWB. Species reactivity: Cow, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-287-099