Learn More
Aviva Systems Biology Corp KRTAP11-1 Antibody - N-terminal region (ARP45923_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45923_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KRTAP11-1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KRTAP11-1. Alternate names: KAP11.1, HACL1, HACL-1ApplicationWB. Species reactivity: Human
Catalog No. 50-014-59168