Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents MSM91076P1
Encompass_Preferred
Product Type: Antibody. Application: ELISA Flow, IF, WB, IHC. Conjugate/Tag: Unconjugated. Clone: KRTH/1076. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Skin, Adenocarcinomas. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human KRT76 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 67kDa (CK1); 64kDa (CK3); 59kDa (CK4); 58kDa (CK5); 56kDa (CK6); 52kDa (CK8).Chromosome: 12q13.13. Gene Symbol: KRT76. Gene ID: 51350. Ensemble: ENSG00000185069. Uniprot: NP_056932.
Catalog No. 50-188-5480