Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents MSM3X446P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, IHC. Conjugate/Tag: Unconjugated. Clone: SPM591. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: PC12 cells, Skin, Prostate carcinoma. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Solubilized keratin extract from human stratum corneum. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 67kDa (CK1); 58kDa (CK5); 56.5kDa (CK10); 50kDa (CK14).Chromosome: Multiple. Gene Symbol: Multiple. Gene ID: 3861. Ensemble: ENSG00000186847. Uniprot: NP_000517.
Catalog No. 50-188-5467