Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents MSM15457P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, IHC. Conjugate/Tag: Unconjugated. Clone: KRT/457. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1. Positive Control: A431 cells, Skin, Colon carcinoma. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Keratin-enriched preparation from cultured human epithelial cells. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: Multiple.Chromosome: Multiple. Gene Symbol: Multiple. Gene ID: 3851. Ensemble: ENSG00000170477. Uniprot: NP_002263.
Catalog No. 50-188-5418