Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents MSM17999P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, IHC. Conjugate/Tag: Unconjugated. Clone: PAN-CK (Cocktail). Clonality: Monoclonal. Host: Mouse. Isotype: IgG.’s kappa. Positive Control: Skin, Adeno- or Squamous carcinomas. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Human epidermal keratin. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 40-67kDa (Multiple).Chromosome: 17q21.2 (KRT77) & 12q13.13 (KRT76). Gene Symbol: KRT. Gene ID: 51350. Ensemble: ENSG00000185069. Uniprot: NP_056932.
Catalog No. 50-188-5427