Learn More
Cusabio Technology LLC Kruppel-like factor 4
Catalog No. 5000051234
Encompass_Preferred
The Human Krueppel-like factor 4 (KLF4) ELISA Kit is designed to quantitatively measure KLF4 levels in serum plasma tissue homogenates and cell lysates using a sandwich ELISA method This kit detects KLF4 with a detection range of 18 75 pg/mL to 1200 pg/mL offering high sensitivity down to 4 6 pg/mL ensuring accurate quantification of low-abundance samples The assay requires a modest sample volume of 50-100 L and delivers results within 1-5 hours Measurements are performed at 450 nm following standard ELISA protocols Ideal for research applications this kit provides reliable and reproducible data for studying KLF4 expression in diverse biological matrices supporting both basic research and preclinical investigations
Catalog No. 50-000-51234 Supplier Cusabio Technology LLC Supplier No. CSBEL012394HU
The Human Krueppel-like factor 4 (KLF4) ELISA Kit is designed to quantitatively measure KLF4 levels in serum plasma tissue homogenates and cell lysates using a sandwich ELISA method This kit detects KLF4 with a detection range of 18 75 pg/mL to 1200 pg/mL offering high sensitivity down to 4 6 pg/mL ensuring accurate quantification of low-abundance samples The assay requires a modest sample volume of 50-100 L and delivers results within 1-5 hours Measurements are performed at 450 nm following standard ELISA protocols Ideal for research applications this kit provides reliable and reproducible data for studying KLF4 expression in diverse biological matrices supporting both basic research and preclinical investigations