Learn More
Lauda Brinkmann Lp KRYO 20 BATH LIQUID 10 L DRUM
Catalog No. 50931352
Encompass_Preferred
Bath Liquid Kryo 20 for LAUDA Circulators, Chillers and Thermostats, for use in applications between the temperature range of -20 to 180oC, 10 L Drum This item is non returnable*
Catalog No. 50-931-352 Supplier Lauda Brinkmann Lp Supplier No. LZB 216
Bath Liquid Kryo 20 for LAUDA Circulators, Chillers and Thermostats, for use in applications between the temperature range of -20 to 180oC, 10 L Drum This item is non returnable*