Learn More
Lauda Brinkmann Lp KRYO 20 BATH LIQUID 20L DRUM
Catalog No. 50931353
Encompass_Preferred
Bath Liquid Kryo 20 for LAUDA Circulators, Chillers and Thermostats, for use in applications between the temperature range of -20 to 180oC, 20 L Drum This item is non returnable*
Catalog No. 50-931-353 Supplier Lauda Brinkmann Lp Supplier No. LZB 316
Bath Liquid Kryo 20 for LAUDA Circulators, Chillers and Thermostats, for use in applications between the temperature range of -20 to 180oC, 20 L Drum This item is non returnable*