Learn More
Lauda Brinkmann Lp KRYO 30 BATH LIQUID 10 L DRUM
Catalog No. 50931349
Encompass_Preferred
Bath Liquid Kryo 30 for LAUDA Circulators, Chillers and Thermostats, for use in applications between the temperature range of -30 to 90oC (in LAUDA Closed System Thermostats -20 to 90oC), 10 L Drum This item is non returnable*
Catalog No. 50-931-349 Supplier Lauda Brinkmann Lp Supplier No. LZB 209
Bath Liquid Kryo 30 for LAUDA Circulators, Chillers and Thermostats, for use in applications between the temperature range of -30 to 90oC (in LAUDA Closed System Thermostats -20 to 90oC), 10 L Drum This item is non returnable*