Learn More
Lauda Brinkmann Lp KRYO 30 BATH LIQUID 5 L DRUM
Catalog No. 50931348
Encompass_Preferred
Bath Liquid Kryo 30 for LAUDA Circulators, Chillers and Thermostats, for use in applications between the temperature range of -30 to 90oC (in LAUDA Closed System Thermostats -20 to 90oC), 5 L Drum This item is non returnable*
Catalog No. 50-931-348 Supplier Lauda Brinkmann Lp Supplier No. LZB 109
Bath Liquid Kryo 30 for LAUDA Circulators, Chillers and Thermostats, for use in applications between the temperature range of -30 to 90oC (in LAUDA Closed System Thermostats -20 to 90oC), 5 L Drum This item is non returnable*