Learn More
Lauda Brinkmann Lp KRYO 51 BATH LIQUID 10 L DRUM
Catalog No. 50931343
Encompass_Preferred
Bath Liquid Kryo 51 for LAUDA Circulators, Chillers and Thermostats, for use in applications between the temperature range of -50 to 120oC, 10 L Drum This item is non returnable*
Catalog No. 50-931-343 Supplier Lauda Brinkmann Lp Supplier No. LZB 221
Bath Liquid Kryo 51 for LAUDA Circulators, Chillers and Thermostats, for use in applications between the temperature range of -50 to 120oC, 10 L Drum This item is non returnable*