Learn More
Lauda Brinkmann Lp KRYO 60 BATH LIQUID 10 L DRUM
Catalog No. 50931337
Encompass_Preferred
Bath Liquid Kryo 60 for LAUDA Circulators, Chillers and Thermostats, for use in applications between the temperature range of -60 to 80oC, 10 L Drum This item is non returnable*
Catalog No. 50-931-337 Supplier Lauda Brinkmann Lp Supplier No. LZB 202
Bath Liquid Kryo 60 for LAUDA Circulators, Chillers and Thermostats, for use in applications between the temperature range of -60 to 80oC, 10 L Drum This item is non returnable*