Learn More
Lauda Brinkmann Lp KRYO 60 BATH LIQUID 5 L DRUM
Catalog No. 50931336
Encompass_Preferred
Bath Liquid Kryo 60 for LAUDA Circulators, Chillers and Thermostats, for use in applications between the temperature range of -60 to 80oC, 5 L Drum This item is non returnable*
Catalog No. 50-931-336 Supplier Lauda Brinkmann Lp Supplier No. LZB 102
Bath Liquid Kryo 60 for LAUDA Circulators, Chillers and Thermostats, for use in applications between the temperature range of -60 to 80oC, 5 L Drum This item is non returnable*