Learn More
GENERAL BIOPHYSICS LLC Krypton Gas Analyzer Model A
Catalog No. NC3361686
Encompass
Thermoconductivity Gas Analyzer for Krypton with Onboard PumpApplication: Calibrated for argon gas with onboard pump to take gas samples.Features: Includes data display screen.Accuracy: ±0.5%.
Catalog No. NC3361686 Supplier GENERAL BIOPHYSICS LLC Supplier No. GAKRO2A
Add to Cart
Add to Cart
Request a Quote
Thermoconductivity Gas Analyzer for Krypton with Onboard PumpApplication: Calibrated for argon gas with onboard pump to take gas samples.Features: Includes data display screen.Accuracy: ±0.5%.